3-Bromo-5-fluoroaniline 95%
Dodavatel: Thermo Scientific
|
Varování
|
H61238.03EA
4600
CZK
H61238.03
3-Bromo-5-fluoroaniline 95%
3-Bromo-5-fluoroaniline
Vzorec:
C₆H₅BrFN MW: 190,01 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07779529 CAS číslo: 134168-97-1 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...