Tisk…

4-Phenyl-1-butyne, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL632058-5GEA 4170 CZK
SIAL632058-5G
4-Phenyl-1-butyne, Sigma-Aldrich®
4-Phenyl-1-butyne
Vzorec: C₁₀H₁₀
MW: 130,19 g/mol
MDL Number: MFCD00041667
CAS číslo: 16520-62-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR