Tisk…

1-Bromo-4-(difluoromethyl)benzene, Sigma-Aldrich®

Dodavatel: Merck

SIAL722472-1GEA 0 CZK
SIAL722472-1G
1-Bromo-4-(difluoromethyl)benzene, Sigma-Aldrich®
1-Bromo-4-(difluoromethyl)benzene
Vzorec: C₇H₅BrF₂
MW: 207,02 g/mol
MDL Number: MFCD06657964
CAS číslo: 51776-71-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR