Tisk…

1-Bromo-4-(heptadecafluorooctyl)benzene, Sigma-Aldrich®

Dodavatel: Merck

SIAL40859-1G-FEA 3680 CZK
SIAL40859-1G-F
1-Bromo-4-(heptadecafluorooctyl)benzene, Sigma-Aldrich®
1-Bromo-4-(heptadecafluorooctyl)benzene
Vzorec: C₁₄H₄BrF₁₇
MDL Number: MFCD03791078
CAS číslo: 206560-77-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR