Tisk…

(Phenylethynyl)tributyltin, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

SIAL340359-10GEA 1760 CZK
SIAL340359-10G
(Phenylethynyl)tributyltin, Sigma-Aldrich®
(Phenylethynyl)tributyltin
Vzorec: C₂₀H₃₂Sn
MDL Number: MFCD00075186
CAS číslo: 3757-88-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR