Tisk…

1-Aminopiperidine, Sigma-Aldrich®


M-Clarity™ quality level = MQ100

SIALA75900-25GEA 7860 CZK
SIALA75900-25G SIALA75900-10G SIALA75900-100G
1-Aminopiperidine, Sigma-Aldrich®
1-Aminopiperidine
Vzorec: C₅H₁₂N₂
MW: 100,16 g/mol
MDL Number: MFCD00006489
CAS číslo: 2213-43-6
EINECS: 218-666-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR