2-Methyl-4-(trifluoromethoxy)benzaldehyde ≥97%
|
Varování
|
H33050.06EA
7210
CZK
H33050.06
H33050.03
2-Methyl-4-(trifluoromethoxy)benzaldehyde ≥97%
2-Methyl-4-(trifluoromethoxy)benzaldehyde
Vzorec:
C₉H₇F₃O₂ MW: 204,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07368717 CAS číslo: 886763-07-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...