Tisk…

Potassium pentyltrifluoroborate, Sigma-Aldrich®

Dodavatel: Merck

SIAL718963-1GEA 2470 CZK
SIAL718963-1G
Potassium pentyltrifluoroborate, Sigma-Aldrich®
Potassium pentyltrifluoroborate
Vzorec: C₅H₁₁BF₃K
MW: 192,0719 g/mol
MDL Number: MFCD07368249
CAS číslo: 446065-12-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR