Tisk…

5-Bromo-2-hydroxy-6-picoline, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL593206-1GEA 2600 CZK
SIAL593206-1G
5-Bromo-2-hydroxy-6-picoline, Sigma-Aldrich®
5-Bromo-2-hydroxy-6-picoline
Vzorec: C₆H₆BrNO
MW: 188,02 g/mol
MDL Number: MFCD03427653
CAS číslo: 54923-31-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR