(±)-2-Hydroxyisovaleric acid ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
H60872.03EA
2270
CZK
H60872.03
(±)-2-Hydroxyisovaleric acid ≥98%
(±)-2-Hydroxyisovaleric acid
Vzorec:
C₅H₁₀O₃ MW: 118,13 g/mol Bod tání: 82…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004242 CAS číslo: 4026-18-0 EINECS: 223-697-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...