Tisk…
Hematocrit tubes acc. to Wintrobe
Dodavatel: Hecht
216-0246EA 98.4 CZK
HECH40535010_U
Hematocrit tubes acc. to Wintrobe
Glass, clear.
  • Fused white graduation
  • Double series in different colours white/red
For determination of the volume of blood corpuscles.