Order Entry
Inserts for storage tubes, 2D Barcoded, Matrix®
Inserts for storage tubes, 2D Barcoded, Matrix®
Inserts for storage tubes, 2D Barcoded, Matrix®
  • Catalog No:
  • 479-1344
  • 479-1343
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  •  
    Colour
    Pk
  • Insert for 0,5 ml and 1,0 ml screw top tube caps
    Insert for 0,5 ml and 1,0 ml screw top tube caps
    479-1344
    4476
    Purple
    500
    1 * 500 KS
    1 * 500 KS - 679,00 Kč
    679,00 Kč
    1 * 500 KS
       679,00 Kč
     : 4476
     : MATR4476
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Insert for 0,5 ml and 1,0 ml screw top tube caps
    Insert for 0,5 ml and 1,0 ml screw top tube caps
    479-1343
    4475
    White
    500
    1 * 500 KS
    1 * 500 KS - 991,00 Kč
    991,00 Kč
    1 * 500 KS
       991,00 Kč
     : 4475
     : MATR4475
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact