Two-stage rotary vane pumps are characterised by their high water vapour tolerance and reliability. Due to their design they withstand large amounts of water vapour in the vacuum pump's inlet vapour stream. This 'water vapour tolerance' is the maximum inlet pressure point at which the pump is able to operate with pure water/fluids and at the same time prevents them of condensing inside the pump.
- Compact design (flexible handle and low weight)
- Low sound level on daily operations
Used widely in vacuum oven, backing pump for high vacuum systems, industrial applications, vacuum manifold (Schlenk line), freeze drying (lyophilisation), glove boxes, vacuum concentrator, HVAC.
Informace o dodávce: Supplied with anti-suck back valve, oil charging, gas ballast valve, centering ring and clamping ring for easy installation and motor overload protection.