Tisk…
Protective hand foam, Schaum-A-Derm
Dodavatel: Kohler
Schaum-A-Derm
115-3249EA 776 CZK
115-3249
Protective hand foam, Schaum-A-Derm
Péče o pokožku Krémy pro pokožku Krémy na ruce
The skin protection foam protects the skin over a period of four hours against substances such as cleaning agents and detergents, organic solvents, acids and caustics, paints and varnishes. Rub a walnut-sized amount onto dry, clean hands and allow to absorb for only two minutes. The unit provides approx. 150 applications.
  • Maintains the skin's sense of touch and ability to grip
  • Forms a thin, viscoelastic, non-stick film on the skin
  • Skin continues to breathe and transpire naturally
  • Dermatologically harmless, preferred by allergy sufferers
Upozornení: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904