These ambidextrous silicone-free gloves are produced in a controlled environment. They feature exceptionally low levels of extractables and particles for excellent product protection.
- Non sterile, powder-free, textured fingertips
- Free of latex proteins, preventing the risk of allergies
- Excellent puncture and chemical resistance
- Optimal electrical properties
Suitable for use in Class 100, ISO 5 or M3.5 cleanrooms.
- Free of latex proteins, preventing the risk of allergies
- Excellent puncture and chemical resistance
- Optimal electrical properties
Ideal for a wide range of industrial sectors, from electronics to paint shops in automotive manufacturing and biotechnologies.
Purity: <2400 particles/cm²
Colour: White
Certifikace: EN ISO 374-1:2016 Type B (JKT), EN ISO 374-5:2016 VIRUS, Cat. III; AQL 2,5
EN 374, Cat. III; AQL 2.5
Balení: Packaging: Nitrilite® gloves are double-wrapped in cleanroom-compatible packaging and packed in a lined shipping carton. 10 master bags per lined carton. 1,000 gloves per case. All packaging and labeling is printed with IPA-resistant ink. The gloves are doublebagged for superior contamination control. Packed in a Class 100 cleanroom.
Balení: 50 gloves per polybag, 2 polybags per master polybag, 10 master polybags per carton. 1,000 gloves per case. All packaging and labeling is printed with IPA resistant ink. The gloves are double bagged for superior contamination control. Packed in a Class 100 cleanroom.