Tisk…
Cleanroom coats, BioClean-R™ Albany
Dodavatel: Ansell-Nitritex
NITRALB001-B20-44EA 0 CZK
NITRALB001-B20-44 NITRALB001-B20-42 NITRALB001-B20-40
Cleanroom coats, BioClean-R™ Albany
These cleanroom coats are made of MT031 fabric consisting of 99% polyester and 1% carbon.
  • Raglan sleeves
  • Centre fastening zip with nickel-free slider
  • Mandarin collar
  • Stud wrist fastenings
The coats are suitable for use in ISO class 4 and above cleanrooms.
Colour: Blue
Weight: 109 g/m²