Tisk…
Micro reaction vials, with cap, Supelco®
Dodavatel: Merck
SIAL33291EA 2850 CZK
SIAL33291
Micro reaction vials, with cap, Supelco®
Micro reaction screw top vessels made of heavy wall borosilicate glass with cone shaped interior for easy removal of small samples.
  • Graduated and clear glass with hole cap
  • Can be autoclaved or centrifuged
Informace o dodávce: Supplied with PTFE red rubber septum and cap.