Tisk…
MCB Glutathione detection kit
Dodavatel: Biotium
30019.EA 12690 CZK
BTIU30019
MCB Glutathione detection kit
Diminished cellular glutathione (GSH) level occurs early in apoptosis due to GSH efflux from mitochondria. Monochlorobimane (MCB) reacts with thiols to form a blue fluorescent product allowing fluorometric quantitation of GSH in cell lysates.