Tisk…
Disposable syringes, 2-piece
Dodavatel: BD Medical
BD™
SEMA1799EA 472 CZK
SEMA1799
Disposable syringes, 2-piece
Stříkačky Střílkačky zdravotnické
Disposable syringe is made of PP, with pistons made of hard PE.
  • Tightness and sterility
  • Pyrogen-free
The pistons run very precisely.
Certifikace: Complies with ISO standards 7886.