Tisk…
Tube rotators, Clifton RM series
Dodavatel: Nickel-Electro
NICKRM-2EA 24920 CZK
NICKRM-2
Tube rotators, Clifton RM series
Třepačky Třepačky rotační
Clifton tube rotators have a small footprint and are ideal for general use. Available as fixed or variable speed models and with hygenic coated body which actively inhibits bacterial growth.
  • Angle of the disc is fully adjustable from vertical to horizontal
  • Benchtop or wall mountable, saving space
  • Illuminated on/off switch
  • Can be used at 4 to 60 °C, in an incubator or cold room
Informace pro objednávání: Range of optional accessory heads available for mixing micro tubes, tubes and flasks.