Tisk…
Magnetic stirring bar retrievers, flexible, PTFE
Dodavatel: Cowie
COWI004.330FEA 2660 CZK
COWI004.330F
Magnetic stirring bar retrievers, flexible, PTFE
Míchadla Nástroje na míchání
Retrivers are used for the removal of stirrer magnets from vessels of all kinds.