Extraction hood, SICCO
Supplier: Bohlender
Certificates
About this item
Anodized aluminium frame with panels made of polycarbonate.
- Transparent and shatter-proof panels
- Robust and light-weight aluminium construction
- Standard connection DN 160 for aspirator tube
- Temperature resistance: from −20 to +70 °C
Top panel with integrated connector for exhaust system offers a good chemical resistance. These extractor hoods are used for a diverse range of industrial applications such as handling of varnish and combustible liquids.
Product Information
- Catalog #Supplier #W×D×H ext.W×D×H int.WeightPkAvailabilityPrice
- Extractor hood, opening dimension 545×600 mmExtractor hood, opening dimension 545×600 mmBOHLV1960-02V1960-02900×600×900 mm860×560×880 mm2011 * 1 KS47 310,00 KčSupplier Article Number: V1960-02Local Article Number: BOHLV1960-02Specifications:
Specifications
Environmentally Preferable - Extractor hood, opening dimension 850×600 mmExtractor hood, opening dimension 850×600 mmBOHLV1960-04V1960-041200×600×900 mm1160×560×880 mm2311 * 1 KS58 140,00 KčSupplier Article Number: V1960-04Local Article Number: BOHLV1960-04Specifications: