Tisk…
VWR®, Hot Start Taq PCR Master Mix, 2X
Dodavatel: Avantor
VWR®
1B1408-100RXNEA 0 CZK
1B1408-100RXN
VWR®, Hot Start Taq PCR Master Mix, 2X
Činidla pro nukleové kyseliny End-point PCR Enzymes and Kits
Hot Start Taq Master Mix, 2X is a ready to use reaction cocktail that contains all required components, except primers and template DNA, for routine PCR amplification of DNA fragments up to 4 kb. The included Taq DNA polymerase is inactivated with proprietary monoclonal antibodies. Incubation at ≥94 °C during PCR thermal cycling irreversibly denatures the antibody and releases full Taq DNA polymerase activity.