Tisk…
Cleanroom finger cots, latex, QRP® 7C
Dodavatel: QRP
BSNC100241 BSNC100239
Cleanroom finger cots, latex, QRP® 7C
Návleky na prsty Návleky na prsty latexové
These disposable, rolled finger cots are ideal for situations where EOS/ESD is a concern. Typical applications include avionics, marine/consumer electronics, IC manufacturing, wire bonding, assembly/repair operations, SMT/PWB assembly and discrete component handling.
  • Anti-static, powder-free, textured finish
  • Suitable for use with 'Class II' devices with thresholds above 1000 V
  • Surface resistivity: 5×10¹² ohms/sq
Suitable for use in Class ISO 5, 100 or M3.5 cleanrooms.
Length: 70 mm
Thickness: 0,08 mm
Colour: Pink
Balení: Packed in a stand-up pouch. 720 finger cots per package.