Tisk…
VWR® NMR Sample Tube Caps, PTFE
Dodavatel: VWR
VWR®
634-0566EA 2680 CZK
634-0566
VWR® NMR Sample Tube Caps, PTFE
Uzávěry Víčka na zkumavky
PTFE, white. Caps are machined to exact specifications with smaller gyroradius and more homogeneous mass distribution than disposable caps providing better spinning stability.
They are recommended for experiments at high - ultra high field and those using chloroform as the reference solution. They will only fit our precision tubes due to the tight tolerance.