Tisk…
Large cryogenic containers, LO series
478-0652EA 321480 CZK
VWRI478-0652 VWRI478-0647 VWRI478-0648
Large cryogenic containers, LO series
The LO 2000 series is a unique range of aluminium cryobiological storage freezers, characterised by high quality, low weight, excellent ergonomics, large sample storage capacity and a low cost per sample stored. All the cryogenic storage containers belonging to LO Series allow direct and easy access to the samples, as each container has a large neck and gas spring assisted lid opening lid (except LO2075). These freezers suit all storage applications that demand frequent access and manipulation of the sample and can be equipped with any kind of storage system for straws, cryo-tubes, vials or blood bags.
  • Storage of racks and canisters (straws, cryovials, blood bags)
  • Large opening, light weight aluminium: easy access, easy to use
  • Can be used for storage of samples in liquid, gas or dry phase
LO Series containers are built in aluminium and have a fiber glass neck, which guarantees very low nitrogen consumption and a lower cost per sample stored (compared to a traditional stainless steel cryogenic container).
The LO Series are equipped with 5 standard castors (2 with brakes) They can also be equipped with an electronic CryoController model S170 or S170-10, which is capable of monitoring temperature, liquid level, alarm status and automatic filling.
Certifikace: The vessels are made in France, according to the highest quality levels.
Compliant with European Medical Device Directive 93/42/EEC.