Tisk…
Single wafer tray, H 20 series, 5" (125 mm)
Dodavatel: Entegris
ENTEH204000011415EA 0 CZK
ENTEH204000011415
Single wafer tray, H 20 series, 5" (125 mm)
Manipulace s wafery Zásobníky na Wafery
Wafer handling trays to safely transport and protect silicon wafers.
  • For internal handling of single wafers
  • Conical pocket holds one wafer facedown contacting only the wafer’s edges
  • Tweezer slots on tray bottom ease wafer removal
  • Stackable for efficient use of storage space, either tray on tray or cover on tray
  • Extended lead times apply