Tisk…
Blotting membranes, nitrocellulose, peqGOLD
732-3197EA 7700 CZK
PEQL330793
Blotting membranes, nitrocellulose, peqGOLD
Blotting membrane made of pure cellulose nitrate.
  • High binding capacity for proteins and nucleic acids
  • Ideally suited for transfers of proteins >15 kDa and colony lifting
  • Nitrocellulose supplied without supporting layer
  • Pore diameter: 0,20 µm