Tisk…
FoodChek™ Salmonella kits
710-1607EA 3920 CZK
FCHKFCSM-003E FCHKFCSM-002E FCHKFCSM-001E FCHKFCSM-004
FoodChek™ Salmonella kits
The FoodChek™ Salmonella test is a fast Salmonella pathogen test, delivering accurate results in less than eight hours including the sample enrichment bacteria amplification or growth phase.
  • Easy-to-use
  • Accordung GLP
  • Cost-effective
  • Profitable and confidence