Tisk…
Grid sieves
Dodavatel: Endecotts
ENDEGRID-31.50EA 0 CZK
ENDEGRID-31.50 ENDEGRID-10.00 ENDEGRID-20.00 ENDEGRID-3.15 ENDEGRID-8.00 ENDEGRID-16.00 ENDEGRID-40.00 ENDEGRID-6.30 ENDEGRID-12.50 ENDEGRID-25.00 ENDEGRID-4.00 ENDEGRID-2.50
Grid sieves
Stainless steel, EN 933-3:1997. Grid sieves are used to determine the flakiness index of aggregates.
  • Sieves with dimensions of 300×300 mm
  • Strong, durable and anti-corrosive