Mobile storage cabinets
Supplier: KOETTERMANN
Certificates
About this item
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Product Information
- Catalog #Supplier #DoorsW×D×HPkAvailabilityPrice
- Mobile storage cabinet with 1 shelfMobile storage cabinet with 1 shelfKOTT315-00057315-00057With front open600×500×590 mm1Log in for availability1 * 1 KS11 070,00 KčSupplier Article Number: 315-00057Local Article Number: KOTT315-00057Specifications:
Specifications
Environmentally Preferable - Mobile storage cabinet, with 2 drawers of 150 mm height and 1 drawer of 300 mm height, lockableMobile storage cabinet, with 2 drawers of 150 mm height and 1 drawer of 300 mm height, lockableKOTT315-00058315-000583 drawers600×516×590 mm1Log in for availability1 * 1 KS24 450,00 KčSupplier Article Number: 315-00058Local Article Number: KOTT315-00058Specifications:
Specifications
Environmentally Preferable - Mobile storage cabinetMobile storage cabinetKOTT315-00059315-000591 door, left hinged600×516×590 mm1Log in for availability1 * 1 KS14 930,00 KčSupplier Article Number: 315-00059Local Article Number: KOTT315-00059Specifications:
Specifications
Environmentally Preferable - Mobile storage cabinet, lockableMobile storage cabinet, lockableKOTT315-00060315-000601 door, left hinged600×516×590 mm1Log in for availability1 * 1 KS15 450,00 KčSupplier Article Number: 315-00060Local Article Number: KOTT315-00060Specifications:
Specifications
Environmentally Preferable - Mobile storage cabinetMobile storage cabinetKOTT315-00061315-000611 door, right hinged600×516×590 mm1Log in for availability1 * 1 KS14 680,00 KčSupplier Article Number: 315-00061Local Article Number: KOTT315-00061Specifications:
Specifications
Environmentally Preferable - Mobile storage cabinetMobile storage cabinetKOTT315-00062315-000621 door, right hinged600×516×590 mm1Log in for availability1 * 1 KS15 390,00 KčSupplier Article Number: 315-00062Local Article Number: KOTT315-00062Specifications:
Specifications
Environmentally Preferable - Mobile storage cabinet, with 3 drawers of 150 mm height, lockableMobile storage cabinet, with 3 drawers of 150 mm height, lockableKOTT315-00063315-000633 drawers600×516×590 mm1Log in for availability1 * 1 KS28 620,00 KčSupplier Article Number: 315-00063Local Article Number: KOTT315-00063Specifications:
Specifications
Environmentally Preferable - Mobile storage cabinet, with 3 drawers of 150 mm height and 1 pencil tray, lockableMobile storage cabinet, with 3 drawers of 150 mm height and 1 pencil tray, lockableKOTT315-00064315-000643 drawers600×516×590 mm1Log in for availability1 * 1 KS36 520,00 KčSupplier Article Number: 315-00064Local Article Number: KOTT315-00064Specifications:
Specifications
Environmentally Preferable - Mobile storage cabinet with 1 shelfMobile storage cabinet with 1 shelfKOTT315-00065315-00065With front open600×500×740 mm1Log in for availability1 * 1 KS12 900,00 KčSupplier Article Number: 315-00065Local Article Number: KOTT315-00065Specifications:
Specifications
Environmentally Preferable - Mobile storage cabinet, with 2 drawers of 300 mm height, lockableMobile storage cabinet, with 2 drawers of 300 mm height, lockableKOTT315-00066315-000662 drawers600×516×740 mm1Log in for availability1 * 1 KS26 170,00 KčSupplier Article Number: 315-00066Local Article Number: KOTT315-00066Specifications:
Specifications
Environmentally Preferable - Mobile storage cabinet with 1 shelfMobile storage cabinet with 1 shelfKOTT315-00067315-000671 door, left hinged600×516×740 mm1Log in for availability1 * 1 KS16 820,00 KčSupplier Article Number: 315-00067Local Article Number: KOTT315-00067Specifications:
Specifications
Environmentally Preferable - Mobile storage cabinet with 1 shelf, lockableMobile storage cabinet with 1 shelf, lockableKOTT315-00068315-000681 door, left hinged600×516×740 mm1Log in for availability1 * 1 KS17 740,00 KčSupplier Article Number: 315-00068Local Article Number: KOTT315-00068Specifications:
Specifications
Environmentally Preferable - Mobile storage cabinet, with 1 drawer of 150 mm heightMobile storage cabinet, with 1 drawer of 150 mm heightKOTT315-00069315-000691 door, left hinged600×516×740 mm1Log in for availability1 * 1 KS20 590,00 KčSupplier Article Number: 315-00069Local Article Number: KOTT315-00069Specifications:
Specifications
Environmentally Preferable - Mobile storage cabinet, with 1 drawer of 150 mm height, lockableMobile storage cabinet, with 1 drawer of 150 mm height, lockableKOTT315-00070315-000701 door, left hinged600×516×740 mm1Log in for availability1 * 1 KS22 890,00 KčSupplier Article Number: 315-00070Local Article Number: KOTT315-00070Specifications:
Specifications
Environmentally Preferable - Mobile storage cabinet with 1 shelfMobile storage cabinet with 1 shelfKOTT315-00071315-000711 door, right hinged600×516×740 mm1Log in for availability1 * 1 KS16 820,00 KčSupplier Article Number: 315-00071Local Article Number: KOTT315-00071Specifications:
Specifications
Environmentally Preferable - Mobile storage cabinet with 1 shelf, lockableMobile storage cabinet with 1 shelf, lockableKOTT315-00072315-000721 door, right hinged600×516×740 mm1Log in for availability1 * 1 KS17 710,00 KčSupplier Article Number: 315-00072Local Article Number: KOTT315-00072Specifications:
Specifications
Environmentally Preferable - Mobile storage cabinet, with 1 drawer of 150 mm heightMobile storage cabinet, with 1 drawer of 150 mm heightKOTT315-00073315-000731 door, right hinged600×516×740 mm1Log in for availability1 * 1 KS20 690,00 KčSupplier Article Number: 315-00073Local Article Number: KOTT315-00073Specifications:
Specifications
Environmentally Preferable - Mobile storage cabinet, with 1 drawer of 150 mm height, lockableMobile storage cabinet, with 1 drawer of 150 mm height, lockableKOTT315-00074315-000741 door, right hinged600×516×740 mm1Log in for availability1 * 1 KS22 830,00 KčSupplier Article Number: 315-00074Local Article Number: KOTT315-00074Specifications:
Specifications
Environmentally Preferable - Mobile storage cabinet, with 2 drawers of 150 mm height and 1 drawer of 300 mm height, lockableMobile storage cabinet, with 2 drawers of 150 mm height and 1 drawer of 300 mm height, lockableKOTT315-00075315-000753 drawers600×516×740 mm1Log in for availability1 * 1 KS30 370,00 KčSupplier Article Number: 315-00075Local Article Number: KOTT315-00075Specifications:
Specifications
Environmentally Preferable - Mobile storage cabinet, lockableMobile storage cabinet, lockableKOTT315-00076315-000764 drawers600×516×740 mm1Log in for availability1 * 1 KS34 560,00 KčSupplier Article Number: 315-00076Local Article Number: KOTT315-00076Specifications: