Tisk…
Scientific calculator
Dodavatel: Texas Instruments
SAFSC6790-1EAEA 720 CZK
SAFSC6790-1EA
Scientific calculator
The power switches automatically in low light conditions either solar and battery with memory safeguard along with wallet style case.
Functions: Math, exponential, trigonometry, inverse trigonometry, statistical functions (mean and standard deviation); angle/time conversions.