Tisk…
Filter holder assembly, All-Glass
Dodavatel: Glassco
GLAS258.202.01EA 6940 CZK
GLAS258.202.01
Filter holder assembly, All-Glass
Zařízení filtrační
Vacuum filter for aqueous, organic and corrosive liquids for particulate contamination analysis. Also recommended for HPLC solvent fi ltration.