Tisk…
Moisturising hand creams, Baktolan® lotion pure
Dodavatel: Bode
Baktolan® Lotion Pure
115-1302EA 208 CZK
115-1302
Moisturising hand creams, Baktolan® lotion pure
Péče o pokožku Krémy pro pokožku Krémy na ruce
Baktolan® lotion pure moisturises and quickly absorbs for daily use on normal skin. Particularly suitable for areas that require staff to continue work quickly.
  • O/W emulsion for normal skin
  • Contains plant-based oils and panthenol
  • Absorbes without leaving a greasy film
  • Colourant- and fragrance-free
Upozornení: * These articles are only available in Germany.
*Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904