Tisk…

Anti-ALB Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
AVIVARP41745_P050EA 11140 CZK
AVIVARP41745_P050
Anti-ALB Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ALB (albumin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR