Tisk…

Anti-CYP11B2 Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
AVIVARP41750_P050EA 11140 CZK
AVIVARP41750_P050
Anti-CYP11B2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CYP11B2 (cytochrome P450, family 11, subfamily B, polypeptide 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR