Tisk…

Anti-F12 Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
AVIVARP41754_P050EA 11140 CZK
AVIVARP41754_P050
Anti-F12 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: F12 (coagulation factor XII (Hageman factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR