Tisk…

Anti-DHODH Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
AVIVARP41941_P050EA 10840 CZK
AVIVARP41941_P050
Anti-DHODH Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: DHODH (dihydroorotate dehydrogenase (quinone))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR