Tisk…

Anti-AMBP Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
AVIVARP41960_P050EA 11140 CZK
AVIVARP41960_P050
Anti-AMBP Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: AMBP (alpha-1-microglobulin/bikunin precursor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Pig, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR