Tisk…
Anti-CRABP2 Rabbit Polyclonal Antibody
Dodavatel: Aviva Systems Biology
AVIVARP41983_P050EA 11140 CZK
AVIVARP41983_P050
Anti-CRABP2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CRABP2 (cellular retinoic acid binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Objednat