Tisk…

Anti-FGF2 Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
AVIVARP42005_P050EA 11140 CZK
AVIVARP42005_P050
Anti-FGF2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: FGF2 (fibroblast growth factor 2 (basic))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR