Tisk…
Anti-GH2 Rabbit Polyclonal Antibody
Dodavatel: Aviva Systems Biology
AVIVARP42013_T100EA 9700 CZK
AVIVARP42013_T100
Anti-GH2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: GH2 (growth hormone 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human