Tisk…

Anti-KRT8 Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
AVIVARP42028_P050EA 11140 CZK
AVIVARP42028_P050
Anti-KRT8 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: KRT8 (keratin 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Pig
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR