Tisk…

Anti-KRT13 Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
AVIVARP42031_T100EA 9430 CZK
AVIVARP42031_T100
Anti-KRT13 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: KRT13 (keratin 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR