Tisk…

Anti-MAP2K3 Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
AVIVARP42064_P050EA 11140 CZK
AVIVARP42064_P050
Anti-MAP2K3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: MAP2K3 (mitogen-activated protein kinase kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR