Tisk…

Anti-PSG5 Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
AVIVARP42067_T100EA 9700 CZK
AVIVARP42067_T100
Anti-PSG5 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PSG5 (pregnancy specific beta-1-glycoprotein 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR