Tisk…

Anti-S100A3 Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
AVIVARP42084_T100EA 9430 CZK
AVIVARP42084_T100
Anti-S100A3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: S100A3 (S100 calcium binding protein A3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR