Tisk…
Anti-SEMG1 Rabbit Polyclonal Antibody
Dodavatel: Aviva Systems Biology
AVIVARP42090_T100EA 9430 CZK
AVIVARP42090_T100
Anti-SEMG1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SEMG1 (semenogelin I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human