Tisk…

Anti-SLC2A5 Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
AVIVARP42096_P050EA 10840 CZK
AVIVARP42096_P050
Anti-SLC2A5 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SLC2A5 (solute carrier family 2 (facilitated glucose/fructose transporter), member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR