Tisk…

Anti-SQLE Rabbit Polyclonal Antibody

Dodavatel: Aviva Systems Biology
AVIVARP42101_P050EA 10840 CZK
AVIVARP42101_P050
Anti-SQLE Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SQLE (squalene epoxidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR